Anti-COPZ1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20440-1-AP-150UL
10085-018EA 402 USD
10085-018
Anti-COPZ1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:K-562 Cells, 1:500-1:5000

Type: Primary
Antigen: COPZ1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR