Anti-INSR Rabbit Polyclonal Antibody

Supplier: Proteintech
20433-1-AP-150UL
10088-710EA 402 USD
10088-710
Anti-INSR Rabbit Polyclonal Antibody
Antibodies
The insulin receptor is a heterotetrameric membrane glycoprotein consisting of disulfide linked α- and β-subunits that are cleaved products of a common precursor.

Western Blot: HEK-293 Cells, 1:500-1:5000

Type: Primary
Antigen: INSR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR