Anti-CXCR7 Rabbit Polyclonal Antibody

Supplier: Proteintech
20423-1-AP-150UL
10085-438EA 402 USD
10085-438
Anti-CXCR7 Rabbit Polyclonal Antibody
Antibodies
CXCR7 (C-X-C chemokine receptor type 7), also known as RDC1, is a member of the G-protein coupled receptor family. CXCR7 can bind the chemokines CXCL11 and CXCL12 with high affinity, and it also acts as coreceptor with CXCR4 for a restricted number of HIV isolates. Expression of CXCR7 has been associated with cardiac development as well as with tumor growth and progression. This antibody recognizes endogenous CXCR7, which has an experimentally determined molecular weight of 50 kDa .

Western Blot: Raji Cells, 1:200-1:2000; IP: HUVEC Cells, 1:200-1:2000

Type: Primary
Antigen: CXCR7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR