Anti-PCNXL2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20417-1-AP-150UL
10091-940EA 402 USD
10091-940
Anti-PCNXL2 Rabbit Polyclonal Antibody
Antibodies
PCNXL2, also named as KIAA0435, belongs to the pecanex family. It may play a role in tumorigenesis of colorectal carcinomas with high microsatellite instability (MSI-H). PCNXL2 is characterized by high mutational frequencies and biallelic mutations in MSI-H colorectal tumors, and is thus likely to be a target gene in these tumors. PCNXL2 is a glycosylation protein. The MW migrates to 260kd.

Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: PCNXL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR