Anti-MIF Rabbit Polyclonal Antibody

Supplier: Proteintech
20415-1-AP-150UL
10090-224EA 402 USD
10090-224
Anti-MIF Rabbit Polyclonal Antibody
Antibodies
Western Blot:Y79 Cells, 1:500-1:5000; IHC: Human lung Tissue, 1:20-1:200

Type: Primary
Antigen: MIF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR