Anti-C11ORF67 Rabbit Polyclonal Antibody

Supplier: Proteintech
20414-1-AP-150UL
10083-754EA 402 USD
10083-754
Anti-C11ORF67 Rabbit Polyclonal Antibody
Antibodies
IHC: Human ovary, 1:20-1:200

Type: Primary
Antigen: C11ORF67
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR