Anti-CCDC158 Rabbit Polyclonal Antibody

Supplier: Proteintech
20396-1-AP-150UL
10084-172EA 402 USD
10084-172
Anti-CCDC158 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Testis Tissue, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200; IP: Mouse Testis Tissue, 1:200-1:2000

Type: Primary
Antigen: CCDC158
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR