Anti-PLEKHF1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20389-1-AP-150UL
10092-462EA 402 USD
10092-462
Anti-PLEKHF1 Rabbit Polyclonal Antibody
Antibodies
PLEKHF1, also named as APPD, LAPF, and ZFYVE15, is a 279 amino acid protein, which contains one PH domain and one FYVE-type zinc finger. PLEKHF1 translocates to lysosome during apoptosis, localizing in the nucleus and cytoplsam. PLEKHF1 is highly expressed in heart and skeletal muscle and weakly expressed in brain, thymus, spleen, kidney, liver, small intestine, placenta and lung. PLEKHF1 may induce apoptosis through the lysosomal-mitochondrial pathway. PLEKHF1 modulates the protein content of the plasma membrane and was involved in different endocytosis events.

Western Blot: Human Skeletal Muscle Tissue, 1:200-1:2000; IF: HepG2 Cells, 1:10-1:100; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: PLEKHF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR