Anti-CLN3 Rabbit Polyclonal Antibody

Supplier: Proteintech
20386-1-AP-150UL
10084-872EA 537.7 USD
10084-872
Anti-CLN3 Rabbit Polyclonal Antibody
Antibodies
Western Blot:SH-SY5Y Cells, 1:200-1:2000; IHC: Human Brain, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: CLN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR