Anti-NPHS2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20384-1-AP-150UL
10091-254EA 402 USD
10091-254
Anti-NPHS2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Colon Tissue, 1:200-1:1000; IF:zebrafish Embryos, 1:20-1:200

Type: Primary
Antigen: NPHS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR