Anti-SFRS2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20371-1-AP-150UL
10094-458EA 402 USD
10094-458
Anti-SFRS2 Rabbit Polyclonal Antibody
Antibodies
SFRS2, also named as PR264 and Splicing component, 35 kDa, belongs to the splicing factor SR family. SFRS2 is necessary for the splicing of pre-mRNA. Some residue of SFRS2 is modified by phosphorylase and acetylase after translation. The molecular weight of modified SFRS2 is 35-40kDa. It is required for formation of the earliest ATP-dependent splicing complex and interacts with spliceosomal components bound to both the 5'- and 3'-splice sites during spliceosome assembly. It also is required for ATP-dependent interactions of both U1 and U2 snRNPs with pre-mRNA. It binds to purine-rich RNA sequences, either 5'-AGSAGAGTA-3' (S=C or G) or 5'-GTTCGAGTA-3'. SFRS2 can bind to beta-globin mRNA and commit it to the splicing pathway. The antibody has no cross reaction to SFRS2B.

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: SFRS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR