Anti-RBBP4 Rabbit Polyclonal Antibody

Supplier: Proteintech
20364-1-AP-150UL
10081-264EA 402 USD
10081-264
Anti-RBBP4 Rabbit Polyclonal Antibody
Antibodies
RBBP4, also named as RBAP48 and CAF-I p48, belongs to the WD repeat RBAP46/RBAP48/MSI1 family. RBBP4 is a core histone-binding subunit that may target chromatin assembly factors, chromatin remodeling factors and histone deacetylases to their histone substrates in a manner that is regulated by nucleosomal DNA. It is a component of several complexes which regulate chromatin metabolism. The antibody is specific to RBBP4.

Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human cervical cancer Tissue, 1:20-1:200

Type: Primary
Antigen: RBBP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR