Anti-UBN1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20363-1-AP-150UL
10096-680EA 402 USD
10096-680
Anti-UBN1 Rabbit Polyclonal Antibody
Antibodies
UBN1, also named as Protein VT4, belongs to the ubinuclein family. UBN1 acts as a novel regulator of senescence. It is involved in the formation of senescence-associated heterochromatin foci (SAHF), which represses expression of proliferation-promoting genes. UBN1 binds to proliferation-promoting genes. It may be required for replication-independent chromatin assembly. The antibody is specific to UBN1.

Western Blot: Mouse Liver Tissue, 1:200-1:2000; IHC: Human cervical cancer, 1:20-1:200

Type: Primary
Antigen: UBN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR