Anti-MYOM1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20360-1-AP-150UL
10090-766EA 537.7 USD
10090-766
Anti-MYOM1 Rabbit Polyclonal Antibody
Antibodies
MYOM1 is a major component of the vertebrate myofibrillar M band. MYOM1 binds myosin, titin, and light meromyosin. This binding is dose dependent. The antibody is specific to MYOM1.

Western Blot: Mouse Skeletal Muscle Tissue, 1:200-1:2000; IHC: Human Skeletal Muscle Tissue, 1:20-1:200

Type: Primary
Antigen: MYOM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR