Anti-PRKAR1A Rabbit Polyclonal Antibody

Supplier: Proteintech
20358-1-AP-150UL
10092-316EA 402 USD
10092-316
Anti-PRKAR1A Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: PRKAR1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR