Anti-TRIM2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20356-1-AP-150UL
10096-320EA 402 USD
10096-320
Anti-TRIM2 Rabbit Polyclonal Antibody
Antibodies
Tripartite motif-containing 2(TRIM2) is an E3 ubiquitin ligase which directs proteasome-mediated degradation of target proteins by the ubiquitination of NEFL and phosphorylation of BCL2L11. This protein contains an N-terminal RING domain, followed by a B-box-2 domain, a coiled-coil region, and 6 C-terminal NHL repeats. TRIM2 involves in the apoptotic response to ischemia via directing degradation of BIM protein, thus has a role in neuroprotection.

Western Blot: Mouse small intestine Tissue, 1:1000-1:10000; IF: Hela Cells, 1:10-1:100; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: TRIM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR