Anti-APLNR Rabbit Polyclonal Antibody

Supplier: Proteintech
20341-1-AP-150UL
10082-998EA 402 USD
10082-998
Anti-APLNR Rabbit Polyclonal Antibody
Antibodies
APLNR, also named as AGTRL1 and APJ, belongs to the G-protein coupled receptor 1 family. It is a receptor for apelin coupled to G proteins that inhibit adenylate cyclase activity. APLNR is an alternative coreceptor with CD4 for HIV-1 infection. It may be involved in the development of AIDS dementia. APLNR is expressed abundantly in the corpus callosum and spinal cord.

  • Western Blot:SH-SY5Y Cells, 1:200-1:2000; IHC: Human Colon Tissue, 1:20-1:200

Type: Primary
Antigen: APLNR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR