Anti-AQP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20333-1-AP-150UL
10083-040EA 402 USD
10083-040
Anti-AQP1 Rabbit Polyclonal Antibody
Antibodies
AQP1 is a member of aquaporins (AQPs) thar are small membrane-spanning proteins facilitating water transport. AQP1 is expressed in most tissues in the mammalian body. Alterations of AQP1 expression have been linked to variety of diseases, including cancer. The predicted molecular weight of AQP1 is around 28 kDa, while highly glycosylated form can also be observed around 40-45 kDa. (1530176)

Western Blot: Human Skeletal Muscle Tissue, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: AQP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR