Anti-ADORA1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20332-1-AP-150UL
10081-906EA 402 USD
10081-906
Anti-ADORA1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Eye Tissue, 1:200-1:2000

Type: Primary
Antigen: ADORA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR