Anti-LTBR Rabbit Polyclonal Antibody

Supplier: Proteintech
20331-1-AP-150UL
10089-838EA 402 USD
10089-838
Anti-LTBR Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human hepatocirrhosis Tissue, 1:20-1:200; FC: HEK-293 Cells, N/A

Type: Primary
Antigen: LTBR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR