Anti-SLC13A4 Rabbit Polyclonal Antibody

Supplier: Proteintech
20302-1-AP-150UL
10094-700EA 402 USD
10094-700
Anti-SLC13A4 Rabbit Polyclonal Antibody
Antibodies
SLC13A4 (also known as SUT-1 or NaS2) belongs to the family of Na+-coupled anion transporters and mediates sulfate reabsorption in the high endothelial venules (HEV). Sulfate is an obligate nutrient for fetal development. Highly expressed in placenta, SLC13A4 has been proposed to play important physiological roles in maintaining high maternal serum sulfate levels during pregnancy and mediating sulfate supply to the fetus.

Western Blot: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: SLC13A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR