Anti-RADIL Rabbit Polyclonal Antibody

Supplier: Proteintech
20284-1-AP-150UL
10092-330EA 402 USD
10092-330
Anti-RADIL Rabbit Polyclonal Antibody
Antibodies
Western Blot: HEK-293 Cells, 1:500-1:5000; IF: Hela Cells, 1:10-1:100; IP: HepG2 Cells, 1:200-1:2000

Type: Primary
Antigen: RADIL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR