Anti-FOXH1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20268-1-AP-150UL
10087-118EA 402 USD
10087-118
Anti-FOXH1 Rabbit Polyclonal Antibody
Antibodies
FOXH1, also named as FAST1 and FAST2, is a transcriptional activator. FOXH1 recognizes and binds to the DNA sequence 5'-TGT[GT][GT]ATT-3'. It is required for induction of the goosecoid (GSC) promoter by TGF-beta or activin signaling. It forms a transcriptionally active complex containing FOXH1/SMAD2/SMAD4 on a site on the GSC promoter called TARE (TGF-beta/activin response element). The antibody is specific to FOXH1.

Western Blot: Mouse Liver Tissue, 1:200-1:1000

Type: Primary
Antigen: FOXH1-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR