Anti-SERINC3 Rabbit Polyclonal Antibody

Supplier: Proteintech
20267-1-AP-150UL
10094-344EA 402 USD
10094-344
Anti-SERINC3 Rabbit Polyclonal Antibody
Antibodies
SERINC, also named as DIFF33 and TDE1, belongs to the TDE1 family. SERINC3 may be involved in cellular transformation. The antibody is specific to SERINC3.

Western Blot: Mouse pancreas Tissue, 1:200-1:2000; IHC: Human lung cancer, 1:20-1:200

Type: Primary
Antigen: SERINC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR