Anti-SERINC2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20266-1-AP-150UL
10094-342EA 402 USD
10094-342
Anti-SERINC2 Rabbit Polyclonal Antibody
Antibodies
SERINC2, also named as TDE2L, belongs to the TDE1 family. SERINC2 contains 11 transmembrane domains, similar to amino acid transporters. The antibody is specific to SERINC2.

Western Blot: HepG2 Cells, 1:200-1:2000

Type: Primary
Antigen: SERINC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR