Anti-SERINC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20265-1-AP-150UL
10094-340EA 402 USD
10094-340
Anti-SERINC1 Rabbit Polyclonal Antibody
Antibodies
SERINC1, also named as KIAA1253, TDE1L and TDE2, belongs to the TDE1 family. SERINC1 enhances the incorporation of serine into phosphatidylserine and sphingolipids. The antibody is specific to SERINC1.

Western Blot: HeLa Cells, 1:200-1:1000; IHC: Human Liver cancer, 1:20-1:200

Type: Primary
Antigen: SERINC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR