Anti-PPP1R11 Rabbit Polyclonal Antibody

Supplier: Proteintech
20263-1-AP-150UL
10092-684EA 402 USD
10092-684
Anti-PPP1R11 Rabbit Polyclonal Antibody
Antibodies
PPP1R11, also named as HCGV and TCTE5, is an inhibitor of protein phosphatase 1. The antibody is specific to PPP1R11.

Western Blot: HeLa Cells, 1:200-1:1000

Type: Primary
Antigen: PPP1R11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR