Anti-FBN2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20252-1-AP-150UL
10086-892EA 402 USD
10086-892
Anti-FBN2 Rabbit Polyclonal Antibody
Antibodies
FBN2 belongs to the fibrillin family. Fibrillins are structural components of 10-12 nm extracellular calcium-binding microfibrils which occur either in association with elastin or in elastin-free bundles. Fibrillin-2-containing microfibrils regulate the early process of elastic fiber assembly. Defects in FBN2 are the cause of congenital contractural arachnodactyly (CCA) which also known as Beals syndrome or distal arthrogryposis type 9 (DA9). The antibody is specific to FBN2.

Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human Placenta Tissue, 1:20-1:200

Type: Primary
Antigen: fibrillin 2-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR