Anti-ROCK2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20248-1-AP-150UL
10093-848EA 402 USD
10093-848
Anti-ROCK2 Rabbit Polyclonal Antibody
Antibodies
ROCK2, also named as KIAA0619, p164 ROCK-2 and Rho kinase 2, belongs to the protein kinase superfamily and AGC Ser/Thr protein kinase family. It is regulates the assembly of the actin cytoskeleton. ROCK2 promotes formation of stress fibers and of focal adhesion complexes. It plays a role in smooth muscle contraction. ROCK2 catalyzes the reaction: ATP + a protein = ADP + a phosphoprotein. The antibody is specific to ROCK2.

Western Blot: HeLa Cells, 1:200-1:1000; IHC: Human Colon, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: ROCK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR