Anti-ROCK1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20247-1-AP-150UL
10093-844EA 402 USD
10093-844
Anti-ROCK1 Rabbit Polyclonal Antibody
Antibodies
ROCK1, also named as p160ROCK and NY-REN-35, belongs to the protein kinase superfamily and AGC Ser/Thr protein kinase family. ROCK1 phosphorylates and activates DAPK3, which then regulates myosin light chain phosphatase through phosphorylation of MYPT1 thereby regulating the assembly of the actin cytoskeleton, cell migration, invasiveness of tumor cells, smooth muscle contraction and neurite outgrowth. ROCK1 is required for centromere positioning and centromere-dependent exit from mitosis. It is necessary for apoptotic membrane blebbing. ROCK1 catalyzes the reaction: ATP + a protein = ADP + a phosphoprotein. The antibody is specific to ROCK1.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human spleen Tissue, 1:20-1:200

Type: Primary
Antigen: ROCK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR