Anti-Neogenin-Specific Rabbit Polyclonal Antibody

Supplier: Proteintech
20246-1-AP-150UL
10091-036EA 402 USD
10091-036
Anti-Neogenin-Specific Rabbit Polyclonal Antibody
Antibodies
NEO1, also named as IGDCC2 and NGN, belongs to the immunoglobulin superfamily and DCC family. NEO1 may be involved as a regulatory protein in the transition of undifferentiated proliferating cells to their differentiated state. NEO1 may also function as a cell adhesion molecule in a broad spectrum of embryonic and adult tissues. The antibody is special to NEO1.

Western Blot: Human Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: Neogenin-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR