Anti-YES1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20243-1-AP-150UL
10097-206EA 402 USD
10097-206
Anti-YES1 Rabbit Polyclonal Antibody
Antibodies
YES1, also named as p61-Yes and YES, belongs to the protein kinase superfamily, Tyr protein kinase family and SRC subfamily. It promotes infectivity of Neisseria gonorrhoeae in epithelial cells by phosphorylating MCP/CD46. YES1 catalyzes the reaction: ATP + a [protein]-L-tyrosine = ADP + a [protein]-L-tyrosine phosphate. The antibody is specific to YES1.

Western Blot: Human Placenta Tissue, 1:200-1:2000; IHC: Human Colon, 1:20-1:200

Type: Primary
Antigen: YES1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR