Anti-UNC5C Rabbit Polyclonal Antibody

Supplier: Proteintech
20240-1-AP-150UL
10096-790EA 402 USD
10096-790
Anti-UNC5C Rabbit Polyclonal Antibody
Antibodies
UNC5C, also named as UNC5H3, belongs to the unc-5 family. It is a receptor for netrin required for axon guidance. It mediates axon repulsion of neuronal growth cones in the developing nervous system upon ligand binding. Axon repulsion in growth cones may be caused by its association with DCC that may trigger signaling for repulsion. It also involved in corticospinal tract axon guidances independently of DCC. It also acts as a dependence receptor required for apoptosis induction when not associated with netrin ligand. The antibody is specific to UNC5C.

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: UNC5C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR