Anti-UNC5A Rabbit Polyclonal Antibody

Supplier: Proteintech
20239-1-AP-150UL
10096-788EA 402 USD
10096-788
Anti-UNC5A Rabbit Polyclonal Antibody
Antibodies
UNC5A, also named as KIAA1976 and UNC5H1, belongs to the unc-5 family. It is a receptor for netrin required for axon guidance. It mediates axon repulsion of neuronal growth cones in the developing nervous system upon ligand binding. Axon repulsion in growth cones may be caused by its association with DCC that may trigger signaling for repulsion. It also acts as a dependence receptor required for apoptosis induction when not associated with netrin ligand. The antibody is specific to UNC5A. It has no cross reaction to UNC5B, UNC5C and UNC5D.

Western Blot: Mouse Brain Tissue, 1:200-1:1000

Type: Primary
Antigen: UNC5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR