Anti-Casein Kinase 2 beta Rabbit Polyclonal Antibody

Supplier: Proteintech
20234-1-AP-150UL
10082-064EA 402 USD
10082-064
Anti-Casein Kinase 2 beta Rabbit Polyclonal Antibody
Antibodies
CSNK2B, also named as CK2N, G5A nd CK II beta, belongs to the casein kinase 2 subunit beta family. CSNK2B participates in Wnt signaling. It plays a complex role in regulating the basal catalytic activity of the alpha subunit. The antibody is specific to CSNK2B.

Western Blot:A431 Cells, 1:200-1:1000

Type: Primary
Antigen: [delete]
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR