Anti-TOP2A Rabbit Polyclonal Antibody

Supplier: Proteintech
20233-1-AP-150UL
10096-198EA 402 USD
10096-198
Anti-TOP2A Rabbit Polyclonal Antibody
Antibodies
TOP2A, also named as TOP2, belongs to the type II topoisomerase family. Topoisomerase II makes double-strand breaks. TOP2 control the topological states of DNA by transient breakage and subsequent rejoining of DNA strands. The antibody is specific to the gene TOP2A.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human spleen, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: TOP2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR