Anti-DLL1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20230-1-AP-150UL
10085-904EA 402 USD
10085-904
Anti-DLL1 Rabbit Polyclonal Antibody
Antibodies
DLL1, also named as Delta1, acts as a ligand for Notch receptors. DLL1 blocks the differentiation of progenitor cells into the B-cell lineage while promoting the emergence of a population of cells with the characteristics of a T-cell/NK-cell precursor. The antibody is specific to the protein DLL1.

Western Blot: Human Placenta Tissue, 1:200-1:2000

Type: Primary
Antigen: DLL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR