Anti-RAB5A Rabbit Polyclonal Antibody

Supplier: Proteintech
20228-1-AP-150UL
10093-216EA 402 USD
10093-216
Anti-RAB5A Rabbit Polyclonal Antibody
Antibodies
RAB5A belongs to the small GTPase superfamily and Rab family. It is required for the fusion of plasma membranes and early endosomes. The antibody is specific to RAB5A. It has no cross reaction to RAB5B and RAB5C.

Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human Placenta Tissue, 1:20-1:200

Type: Primary
Antigen: RAB5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR