Anti-ROBO4 Rabbit Polyclonal Antibody

Supplier: Proteintech
20221-1-AP-150UL
10093-840EA 402 USD
10093-840
Anti-ROBO4 Rabbit Polyclonal Antibody
Antibodies
ROBO4 belongs to the immunoglobulin superfamily and ROBO family. It is a receptor for Slit proteins, at least for SLIT2, and seems to be involved in angiogenesis and vascular patterning. ROBO4 may mediate the inhibition of primary endothelial cell migration by Slit proteins. The antibody is special to ROBO4.

Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: ROBO4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR