Anti-ROBO1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20219-1-AP-150UL
10093-834EA 524.48 USD
10093-834
Anti-ROBO1 Rabbit Polyclonal Antibody
Antibodies
ROBO1, also named as DUTT1, belongs to the immunoglobulin superfamily and ROBO family. It is a receptor for SLIT1 and SLIT2 which are thought to act as molecular guidance cue in cellular migration, including axonal navigation at the ventral midline of the neural tube and projection of axons to different regions during neuronal development. In axon growth cones, the silencing of the attractive effect of NTN1 by SLIT2 may require the formation of a ROBO1-DCC complex. It may be required for lung development. The antibody is specific to ROBO1.

Western Blot: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: ROBO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR