Anti-IGF1B Rabbit Polyclonal Antibody

Supplier: Proteintech
20215-1-AP-150UL
10088-648EA 402 USD
10088-648
Anti-IGF1B Rabbit Polyclonal Antibody
Antibodies
IGF1B, also named as IBP1, MGF, IGF-I and Somatomedin-C, belongs to the insulin family. IGF1 is structurally and functionally related to insulin but have a much higher growth-promoting activity. Altered expression or mutation of IGF-1 is associated with several human disorders, including type I diabetes and various forms of cancer. Defects in IGF1 are the cause of insulin-like growth factor I deficiency (IGF1 deficiency) which is an autosomal recessive disorder characterized by growth retardation, sensorineural deafness and mental retardation. The antibody is specific to isoform IGF-1B.

Western Blot:PC-3 Cells, 1:500-1:5000; IHC: Human Liver cancer, 1:20-1:200

Type: Primary
Antigen: IGF1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR