Anti-GPRIN3 Rabbit Polyclonal Antibody

Supplier: Proteintech
20212-1-AP-150UL
10087-808EA 402 USD
10087-808
Anti-GPRIN3 Rabbit Polyclonal Antibody
Antibodies
GPRIN3, also named as KIAA2027, may be involved in neurite outgrowth. The antibody is specific to GPRIN3.

Western Blot: Human Brain Tissue, 1:200-1:2000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: GPRIN3-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR