Anti-NARFL Rabbit Polyclonal Antibody

Supplier: Proteintech
20209-1-AP-150UL
10090-816EA 402 USD
10090-816
Anti-NARFL Rabbit Polyclonal Antibody
Antibodies
NARFL, also named as PRN and IOP1, belongs to the NARF family. NAFL is a component of the cytosolic iron-sulfur (Fe/S) protein assembly machinery. It is required for maturation of extramitochondrial Fe/S proteins. It seems to negatively regulate the level of HIF1A expression, although this effect could be indirect. [18270200], [16956324] The antibody is specific to NARFL.

Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human Kidney, 1:20-1:200

Type: Primary
Antigen: NARFL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR