Anti-POTED Rabbit Polyclonal Antibody

Supplier: Proteintech
20206-1-AP-150UL
10092-604EA 402 USD
10092-604
Anti-POTED Rabbit Polyclonal Antibody
Antibodies
POTED, also named as A26B3, ANKRD21 and POTE, belongs to the POTE family. POTE has an important signaling function in the reproductive system. The antibody is specific to POTED.

Western Blot: Mouse Testis Tissue, 1:200-1:2000

Type: Primary
Antigen: POTED
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR