Anti-POTEA Rabbit Polyclonal Antibody

Supplier: Proteintech
20203-1-AP-150UL
10092-602EA 402 USD
10092-602
Anti-POTEA Rabbit Polyclonal Antibody
Antibodies
POTEA, also named as A26A1 and POTE8, belongs to the POTE family. POTE has an important signaling function in the reproductive system. The antibody is specific to POTEA.

IHC: Human ovary, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: POTEA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR