Anti-STEAP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
20201-1-AP-150UL
10095-290EA 402 USD
10095-290
Anti-STEAP2 Rabbit Polyclonal Antibody
Antibodies
STEAP2, also named as PCANAP1 and STAMP1, belongs to the STEAP family. It is a metalloreductase that has the ability to reduce both Fe3+ to Fe2+ and Cu2+ to Cu1+. STEAP2 uses NAD+ as acceptor. The antibody is specific to STEAP2.

Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human Heart Tissue, 1:20-1:200

Type: Primary
Antigen: STEAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR