Anti-STEAP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
20199-1-AP-150UL
10095-288EA 537.7 USD
10095-288
Anti-STEAP1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A431 Cells, 1:200-1:2000

Type: Primary
Antigen: STEAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR