Anti-BIN3 Rabbit Polyclonal Antibody

Supplier: Proteintech
20186-1-AP-150UL
10083-610EA 402 USD
10083-610
Anti-BIN3 Rabbit Polyclonal Antibody
Antibodies
BIN3, also named as MEPCE and BCDIN3, belongs to the methyltransferase superfamily. It is an S-adenosyl-L-methionine-dependent methyltransferase that adds a methylphosphate cap at the 5'-end of 7SK snRNA, leading to stabilize it.

Western Blot:Jurkat Cells, 1:200-1:2000

Type: Primary
Antigen: BIN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR