Anti-TRAPPC9 Rabbit Polyclonal Antibody

Supplier: Proteintech
20176-1-AP-150UL
10081-186EA 402 USD
10081-186
Anti-TRAPPC9 Rabbit Polyclonal Antibody
Antibodies
TRAPPC9, also named as KIAA1882 and NIBP, belongs to the NIBP family. It functions as an activator of NF-kappa-B through increased phosphorylation of the IKK complex. TRAPPC9 may function in neuronal cells differentiation. It may play a role in vesicular transport from endoplasmic reticulum to Golgi. TRAPPC9 was found in neurons of the cerebral cortex, hippocampus, and deep gray matter. In WB, the MW of TRAPPC9 is 130-140kd. A 250kd maybe the TRAPPC9 undergoes extensive posttranslational modification.

Western Blot:Neuro-2a Cells, 1:200-1:2000

Type: Primary
Antigen: TRAPPC9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR