Anti-NCOA5 Rabbit Polyclonal Antibody

Supplier: Proteintech
20175-1-AP-150UL
10090-880EA 537.7 USD
10090-880
Anti-NCOA5 Rabbit Polyclonal Antibody
Antibodies
Nuclear receptor coactivator 5 (NCOA5), also known as coactivator independent of AF-2 (CIA), possesses both repressor and activator functions . It can interact with Estrogen Receptor (ER) alpha and ER beta in a ligand-dependent manner. NCOA5 has been shown to regulate the ER alpha-mediated transcription as well as the expression of the c-Myc gene . Recently identified as a conserved component of pluripotent stem cells, mammalian NCOA5 may contribute to the maintenance of the pluripotent stem cell population .

Western Blot: HEK-293 Cells, 1:500-1:5000; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: NCOA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR